Please login to your online account to display your discounted pricing

Bromine-1,4-dioxane complex, 0.25M solution in 1,4-dioxane, AcroSeal™, ACROS Organics™

$140.81 - $140.81

Chemical Identifiers

Chemical Name or Material Bromine-1, 4-dioxane complex
Name Note 0.25M solution in 1, 4-dioxane
CAS 15481-39-7
CAS 123-91-1
Molecular Formula C4H8Br2O2
Formula Weight 247.91
MDL Number MFCD00034818
Synonym Dioxane dibromide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388411000

  SDS
thermo fisher scientific
ACAC388411000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $140.81
Description & Specifications

Specifications

Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Health Hazard 2 GHS H Statement
Suspected of causing cancer. Fatal if inhaled. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Causes serious eye irritation. Causes skin irritation. Highly flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.
Physical Form Liquid
Formula Weight 247.91
Quantity 100mL
Color Clear red
Packaging AcroSeal™ Glass Bottle in Tin Can
Concentration 0.2 to 0.3M
CAS 123-91-1
CAS 15481-39-7
Synonym Dioxane dibromide
Name Note 0.25M solution in 1, 4-dioxane
Molecular Formula C4H8Br2O2
Chemical Name or Material Bromine-1, 4-dioxane complex
MDL Number MFCD00034818