missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromide, Ion chromatography standard solution, Specpure™, Br- 1000μg/mL

$157.00 - $157.00

Chemical Identifiers

MDL Number MFCD00134172
Chemical Name or Material Bromide Ion chromatography standard solution, Br- 1000μg/mL
Grade Specpure™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35552AE SDS Alfa Aesar™
35552AE
100mL Each for $157.00
AA35552AP SDS Alfa Aesar™
35552AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00134172
Bromide Ion chromatography standard solution, Br- 1000μg/mL
Specpure™

Specifications

Liquid
100mL
Specpure™
MFCD00134172
Bromide Ion chromatography standard solution, Br- 1000μg/mL

Safety and Handling

TSCA : TSCA