missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromhexine hydrochloride, 98+%

A mucolytic agent

$147.27 - $479.59

Chemical Identifiers

CAS 611-75-6
Molecular Formula C14H21Br2ClN2
Molecular Weight (g/mol) 412.594
MDL Number MFCD00056626
InChI Key UCDKONUHZNTQPY-UHFFFAOYSA-N
Synonym bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
PubChem CID 5702220
ChEBI CHEBI:31303
IUPAC Name 2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]aniline;hydrochloride
SMILES CN(CC1=CC(=CC(=C1N)Br)Br)C2CCCCC2.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6345122 View Documents Alfa Aesar™
J6345122
100g Each for $147.27
 
AAJ6345136 View Documents Alfa Aesar™
J6345136
500g Each for $479.59
 
Specifications

Chemical Identifiers

611-75-6
412.594
UCDKONUHZNTQPY-UHFFFAOYSA-N
5702220
2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]aniline;hydrochloride
C14H21Br2ClN2
MFCD00056626
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
CHEBI:31303
CN(CC1=CC(=CC(=C1N)Br)Br)C2CCCCC2.Cl

Specifications

White
100g
C14H21Br2ClN2
4848376
bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
CN(CC1=CC(=CC(=C1N)Br)Br)C2CCCCC2.Cl
412.594
CHEBI:31303
Powder
Bromhexine hydrochloride
240°C to 244°C
611-75-6
MFCD00056626
14,1391
UCDKONUHZNTQPY-UHFFFAOYSA-N
2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]aniline;hydrochloride
5702220
412.6
≥98%
Safety and Handling

Safety and Handling

RTECSNumber : XS9950000