Please login to your online account to display your discounted pricing

Bromelain, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC402835000

 View more versions of this product

Catalog No. AC402835000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Purity Grade Extra Pure
Concentration 2.4 to 3 FIP U/mg
Quantity 500g
Color Cream to slightly brown
Merck Index 15, 1395
Assay 2.4 to 3 FIP U/mg
RTECS Number EF8575000
EINECS Number 232-572-4
TSCA TSCA
pH 3 to 5
CAS 9001-00-7
Packaging Glass bottle
Name Note Pure
MDL Number MFCD00081423
Chemical Name or Material Bromelain
Loss on Drying 5% max.
Residue after Ignition 5% max. ()
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material Bromelain
Name Note Pure
CAS 9001-00-7
Purity Grade Extra Pure
Assay 2.4 to 3 FIP U/mg
MDL Number MFCD00081423
Accessories

GHS P Statement:
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents