Learn More
Bromelain, sourced from stem, 1200 GDU/g, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 402831000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bromelain | |
37189-34-7 | |
3 to 5 | |
5% max. | |
Glass bottle | |
100 g | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether | |
Extra Pure | |
Powder |
Pure | |
Brown to Cream | |
5% max. () | |
Authentic | |
MFCD00081423 | |
15, 1395 | |
2.4 to 3 FIP U/mg | |
2.4 to 3 FIP U/mg |
Chemical Identifiers
37189-34-7 | |
MFCD00081423 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 253-387-5
RTECSNumber : EF8575000
TSCA : TSCA
RUO – Research Use Only