missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromelain, sourced from stem, 2500 GDU/g, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448211000
Specifications
Bromelain | |
5% max | |
2500 GDU/g | |
MFCD00081423 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether |
Glass bottle | |
100g | |
37189-34-7 | |
15,1395 |
Chemical Identifiers
37189-34-7 | |
MFCD00081423 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 253-387-5