missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromate-Bromide Solution, 0.0200N, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR116400010F

 View more versions of this product

Catalog No. R116400010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Bromate-Bromide Solution
7758-01-2,7758-01-2
dihydrogen monoxide
Odorless
Solubility in Water: Infinite
Colorless
0.0200 N (N/50)
0.02000 ± 0.00002 N at 20°C
Liquid
Cubitainer
Aqueous Solution
10L
Documents
Promotions

Promotions