Please login to your online account to display your discounted pricing

Bromate-Bromide, For Phenols, Certified, 0.100N ±0.001N (0.017M), LabChem™

$47.66 - $135.91

Chemical Identifiers

Chemical Name or Material Bromate-Bromide
CAS 7732-18-5
CAS Max % 98.72
CAS 7758-02-3 
CAS Max % 1
CAS 7758-01-2 
CAS Max % 0.28
Name Note For Phenols
Purity Grade Certified Grade
Linear Formula KBrO3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC119501

 
lab chem
LC119501
500mL Each for $47.66

LC119502

 
lab chem
LC119502
1L Each for $63.89

LC119504

 
lab chem
LC119504
4L Each for $135.91
Description & Specifications

Specifications

Formula Weight 167
Purity Grade Certified Grade
Decomposition Information Hydrogen bromide
CAS 7758-01-2 
CAS Max % 98.72
Solubility Soluble in water
CAS 7758-02-3 
Health Hazard 1 Warning
Color Clear, colorless
Quantity 500mL
Physical Form Liquid
Linear Formula KBrO3
Storage Information Room Temperature
Identification Passes Test
Health Hazard 2 GHS H Statement
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.
CAS Max % 1
Traceability to NIST Traceable to NIST
CAS Max % 0.28
Concentration 0.100N ±0.001N (0.017M)
Packaging Amber Glass
Chemical Name or Material Bromate-Bromide
Name Note For Phenols
CAS 7732-18-5
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.

GHS H Statement
Suspected of causing cancer.