Please login to your online account to display your discounted pricing

Brock Magiscope™


Simple design of this durable scope yields extraordinarily crisp, bright images.

Manufacturer: fisher scientific  MODEL70

 View more versions of this product

Catalog No. S197094

  • / Each
 

Description & Specifications

Specifications

Warranty Lifetime
Axis Type Brass-on-Brass friction focusing, fine adjustment ramp
Design Type Rubber band stage clip
Eyepiece 5X Huygenian
Magnification Power 20x
Objective Types 4x achromatic
For Use With (Application) Science classroom use
Head Style Monocular
Material Solid Aluminum Alloy
Weight (English) 28.8 oz.
Weight (Metric) 816.46g
Height (Metric) 24.13cm
Includes 4X achromatic objective
Height (English) 9.5 in.