Please login to your online account to display your discounted pricing
Brock Magiscope™
Simple design of this durable scope yields extraordinarily crisp, bright images.
$49.30 - $313.20
Specifications
For Use With (Application) Science classroom use
View More Specs
Products
Catalog Number Mfr. No. Magnification Power Objective Types Price Quantity    
S197094
fisher scientific
MODEL70
20x 4x achromatic Each for $313.20
S19709A
fisher science education
OBJ
50x 10x achromatic Each for $49.30
S19709D
fisher science education
10X
40x 10x widefield Each for $62.10
Description & Specifications
Specifications
For Use With (Application) Science classroom use
Features Lumarod™ fiber-optic illuminator, a unique light-gathering rod that delivers an amazing amount of light to illuminate slides.
  • 5X Huygenian eyepieces
  • With 4X achromatic objectives; 10X available separately (below)
  • Provides 20X magnification
  • Focus by sliding eyetube up and down