Please login to your online account to display your discounted pricing

BRL 54443, Tocris

Potent 5-ht1E/F agonist

$159.50 - $662.25

Chemical Identifiers

Chemical Name or Material BRL 54443
Assay Percent Range >98%
Molecular Formula C14H18N2O
Formula Weight 230.31
Synonym 5-Hydroxy-3-(1-methylpiperidin-4-yl)-1H-indole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

112910

 
r & d systems
112910
10mg Each for $159.50

112950

 
r & d systems
112950
50mg Each for $662.25
Description & Specifications

Specifications

Chemical Name or Material BRL 54443
Quantity 10mg
CAS 57477-39-1
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C14H18N2O
Formula Weight 230.31
Synonym 5-Hydroxy-3-(1-methylpiperidin-4-yl)-1H-indole
Safety and Handling

Desiccate at +4°C