Please login to your online account to display your discounted pricing

BRL 54443, Tocris


Potent 5-ht1E/F agonist

Quantity 50mg

Manufacturer: r & d systems  112950

 View more versions of this product

Catalog No. 112950

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material BRL 54443
Quantity 50mg
CAS 57477-39-1
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C14H18N2O
Formula Weight 230.31
Synonym 5-Hydroxy-3-(1-methylpiperidin-4-yl)-1H-indole

Specifications

Chemical Name or Material BRL 54443
Assay Percent Range >98%
Molecular Formula C14H18N2O
Formula Weight 230.31
Synonym 5-Hydroxy-3-(1-methylpiperidin-4-yl)-1H-indole
Accessories

Desiccate at +4°C