Please login to your online account to display your discounted pricing

BRL 52537 hydrochloride, Tocris

Potent and selective κ agonist

$164.61 - $653.32

Chemical Identifiers

Chemical Name or Material BRL 52537 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H24Cl2N2O·HCl
Formula Weight 391.77
Synonym (±)-1-(3, 4-Dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

069910

 
r & d systems
RDS069910
10mg Each for $164.61

069950

 
r & d systems
RDS069950
50mg Each for $653.32
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 391.77
Chemical Name or Material BRL 52537 hydrochloride
Assay Percent Range >98%
Synonym (±)-1-(3, 4-Dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride
Molecular Formula C18H24Cl2N2O·HCl
CAS 112282-24-3
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature