Please login to your online account to display your discounted pricing

BRL 44408 maleate, Tocris


Selective α2A antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS113310

 View more versions of this product

Catalog No. 113310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 331.37
Quantity 10mg
Chemical Name or Material BRL 44408 maleate
Assay Percent Range >98%
Molecular Formula C13H17N3·C4H4O4
CAS 681806-46-2
Synonym 2-[(4, 5-Dihydro-1H-imidazol-2-yl)methyl]-2, 3-dihydro-1-methyl-1H-isoindole maleate
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material BRL 44408 maleate
Assay Percent Range >98%
Molecular Formula C13H17N3·C4H4O4
Formula Weight 331.37
Synonym 2-[(4, 5-Dihydro-1H-imidazol-2-yl)methyl]-2, 3-dihydro-1-methyl-1H-isoindole maleate
Accessories

Desiccate at -20°C