Please login to your online account to display your discounted pricing

BRL 37344, sodium salt, Tocris


β3 agonist

Quantity 50mg

Manufacturer: r & d systems  RDS094850

 View more versions of this product

Catalog No. 094850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 385.82
Assay Percent Range >98%
Molecular Formula C19H21NO4ClNa
CAS 127299-93-8
Synonym (R™, R™)-(±)-4-[2-[(2-(3-Chlorophenyl)-2-hydroxyethyl)amino]propyl]phenoxyacetic acid, sodium salt
Chemical Name or Material BRL 37344, sodium salt
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material BRL 37344, sodium salt
Assay Percent Range >98%
Molecular Formula C19H21NO4ClNa
Formula Weight 385.82
Synonym (R™, R™)-(±)-4-[2-[(2-(3-Chlorophenyl)-2-hydroxyethyl)amino]propyl]phenoxyacetic acid, sodium salt
Accessories

Store at Room Temperature