Please login to your online account to display your discounted pricing

BRL 37344, sodium salt, Tocris

β3 agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material BRL 37344, sodium salt
Assay Percent Range >98%
Molecular Formula C19H21NO4ClNa
Formula Weight 385.82
Synonym (R™, R™)-(±)-4-[2-[(2-(3-Chlorophenyl)-2-hydroxyethyl)amino]propyl]phenoxyacetic acid, sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

094810

 
r & d systems
RDS094810
10mg Each for $205.70

094850

 
r & d systems
RDS094850
50mg Each for $862.95
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 385.82
Chemical Name or Material BRL 37344, sodium salt
CAS 127299-93-8
Assay Percent Range >98%
Molecular Formula C19H21NO4ClNa
Synonym (R™, R™)-(±)-4-[2-[(2-(3-Chlorophenyl)-2-hydroxyethyl)amino]propyl]phenoxyacetic acid, sodium salt
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature