BRL 15572 hydrochloride, Tocris
Selective h5-HT1D antagonist
Manufacturer:  R & D Systems 120710
 View more versions of this product
Catalog No. 120710
Specifications Description & Specifications
Specifications
BRL 15572 hydrochloride
1173022-77-9
C25H27ClN2O·HCl
3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride
10mg
>98%
443.42
Chemical Identifiers
>98%
443.42
BRL 15572 hydrochloride
C25H27ClN2O·HCl
3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride
Safety and Handling
Recommended Storage : Desiccate at +4°C