Please login to your online account to display your discounted pricing

BRL 15572 hydrochloride, Tocris

Selective h5-HT1D antagonist

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material BRL 15572 hydrochloride
Assay Percent Range >98%
Molecular Formula C25H27ClN2O·HCl
Formula Weight 443.42
Synonym 3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

120750

 
r & d systems
120750
50mg Each for $834.54

120710

 
r & d systems
120710
10mg Each for $199.70
Description & Specifications

Specifications

Formula Weight 443.42
Quantity 50mg
Chemical Name or Material BRL 15572 hydrochloride
Assay Percent Range >98%
Molecular Formula C25H27ClN2O·HCl
CAS 1173022-77-9
Synonym 3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C