Please login to your online account to display your discounted pricing
Metrohm Model 727 Stirring Stand
$203.13 - $203.13
Products
Catalog Number Mfr. No. Price Quantity    
BR20757507
metrohm
020757507
Each for $203.13
Description & Specifications
Titration stand and stirring accessory is available witn magnetic stirrer and electrode rinse or with propeller stirrer. Models for 9VDC connect directly to Metrohm Titrators.