Please login to your online account to display your discounted pricing
Brinkmann™ Bottle-Top Adapters
$55.83 - $69.17
Products
Catalog Number Mfr. No. Price Quantity    
13-688-148
metrohm canada
KITLSADP28S*
Pack of 3 Each for $55.83
13-688-149
metrohm canada
KITLSADP38S*
Pack of 3 Each for $69.17
Description & Specifications
Allow attachment of dispensers to various sizes of bottles. Choose PTFE adapters for use with strong acids, strong alkalis, or aggressive reagents; polypropylene for most other reagents.
‡Pack of three includes one adapter of each diameter.