Please login to your online account to display your discounted pricing

Brilliant Green, Reagent Grade, LabChem™

$77.97 - $77.97

Chemical Identifiers

Chemical Name or Material Brilliant Green
CAS 633-03-4
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C27H33N2·HO4S
Formula Weight 482.65
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC117958

 
lab chem
LC117958
Amber Glass 25g Each for $77.97
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 482.65
Chemical Name or Material Brilliant Green
Quantity 25g
Purity Grade Reagent Grade
Linear Formula C27H33N2·HO4S
CAS 633-03-4
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Melting Point 210°C
Color Gray-green
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

Room Temperature