Please login to your online account to display your discounted pricing

Brilliant Green, HARLECO™, EMD Millipore


Quantity 50g
Packaging Glass Bottle

Manufacturer: emd millipore corporation  16214

 View more versions of this product

Catalog No. M16214

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 482.64
Quantity 50g
Physical Form Powder
Purity Grade Certified Biological Stain
Packaging Glass Bottle
Chemical Name or Material Brilliant Green
Name Note HARLECO™
CAS 633-03-4
Molecular Formula C27H33N2 · HSO4
Synonym Malachite Green G

Specifications

Chemical Name or Material Brilliant Green
Name Note HARLECO™
CAS 633-03-4
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C27H33N2 · HSO4
Formula Weight 482.64
Synonym Malachite Green G