Please login to your online account to display your discounted pricing

Brilliant Green, HARLECO™, EMD Millipore

$318.57 - $455.60

Chemical Identifiers

Chemical Name or Material Brilliant Green
Name Note HARLECO™
CAS 633-03-4
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C27H33N2 · HSO4
Formula Weight 482.64
Synonym Malachite Green G
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M16214

 
emd millipore corporation
16214
Glass Bottle 50g Each for $455.60

M16212

 
emd millipore corporation
16212
Glass Bottle 25g Each for $318.57
Description & Specifications

Specifications

Chemical Name or Material Brilliant Green
Name Note HARLECO™
CAS 633-03-4
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C27H33N2 · HSO4
Formula Weight 482.64
Synonym Malachite Green G
Packaging Glass Bottle
Quantity 50g