Please login to your online account to display your discounted pricing

Brilliant Green, HARLECO™, EMD Millipore


Quantity 25g
Packaging Glass Bottle

Manufacturer: emd millipore corporation  16212

 View more versions of this product

Catalog No. M16212

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 482.64
Quantity 25g
Purity Grade Certified Biological Stain
Physical Form Powder
Chemical Name or Material Brilliant Green
Name Note HARLECO™
Molecular Formula C27H33N2 · HSO4
CAS 633-03-4
Synonym Malachite Green G
Packaging Glass Bottle

Specifications

Chemical Name or Material Brilliant Green
Name Note HARLECO™
CAS 633-03-4
Purity Grade Certified Biological Stain
Physical Form Powder
Molecular Formula C27H33N2 · HSO4
Formula Weight 482.64
Synonym Malachite Green G