Please login to your online account to display your discounted pricing

Brilliant Green, 1% Aqueous, pH 0.0 to 2.6 Yellow to Green, Certified, LabChem™

$96.74 - $96.74

Chemical Identifiers

Chemical Name or Material Brilliant Green
CAS 7732-18-5
CAS Max % 99
CAS 633-03-4
CAS Max % 1
Name Note 1% Aqueous, pH 0.0 to 2.6 Yellow to Green
Purity Grade Certified Grade
Linear Formula C27H33N2·HO4S
Formula Weight 482.65
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC118001

 
lab chem
LC118001
Poly Bottle 500mL Each for $96.74
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 482.65
Name Note 1% Aqueous, pH 0.0 to 2.6 Yellow to Green
CAS 7732-18-5
Chemical Name or Material Brilliant Green
Quantity 500mL
Physical Form Liquid
Linear Formula C27H33N2·HO4S
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 633-03-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
CAS Max % 1
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Dark green
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature