missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Brilliant Crocein

Manufacturer:  Alfa Aesar™ A1464230

 View more versions of this product

Catalog No. AAA1464230



Description

Description

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Brilliant Crocein
C22H16N4Na2O7S2
C.I. 27290; Crocein Scarlet MOO
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
88641610
250g
5413-75-2
MFCD00003907
Soluble in water
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
558.491
556.49

Chemical Identifiers

5413-75-2
558.491
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
88641610
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
C22H16N4Na2O7S2
MFCD00003907
C.I. 27290; Crocein Scarlet MOO
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents