Please login to your online account to display your discounted pricing

Coonmassie Brilliant Blue R-250, Fisher BioReagents


Quantity 50g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP10150

 View more versions of this product

Catalog No. BP101-50

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 825.96
Melting Point 174°C
Quantity 50g
pH 6.2
Molecular Formula C45H44N3NaO7S2
Density 0.25g/cm³
Color Index No. 42660
Color Red Brown
Chemical Name or Material Brilliant Blue R-250
CAS 6104-59-2
ChemAlert Storage Symbol Gray
Identification Pass Test
Synonym Coomassie Blue R-250
Loss on Drying 5% max.
Packaging Amber Glass
Recommended Storage RT
Extinction Coefficient 700g(-1) cm(-1) min.
Lambda Max 590 to 594nm

Specifications

Chemical Name or Material Brilliant Blue R-250
CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Formula Weight 825.96
Synonym Coomassie Blue R-250
Accessories
 

RT

SDS & Safety Documents