missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coomassie Brilliant Blue G-250, Fisher BioReagents

$121.29 - $184.14

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
Molecular Weight (g/mol) 854.025
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Synonym Coomassie™ Blue G-250
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP100-25 SDS View Documents Fisher BioReagents
BP10025
25g Glass Bottle Each for $121.29
Case of 6 Each for $727.74
 
BP100-50 SDS View Documents Fisher BioReagents
FLBP10050
50g Glass Bottle Each for $184.14
Case of 6 Each for $1,104.84
 
Description

Description

Brilliant Blue G-250 is a general protein stain.
Specifications

Chemical Identifiers

6104-58-1
854.025
Coomassie™ Blue G-250
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
C47H48N3NaO7S2
RWVGQQGBQSJDQV-UHFFFAOYSA-M
6328534
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

6104-58-1
C47H48N3NaO7S2
Pass Test
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
854.025
6328534
854.02
Glass Bottle
Blue
Brilliant Blue G-250
600g(-1) cm(-1) min.
Coomassie™ Blue G-250
RWVGQQGBQSJDQV-UHFFFAOYSA-M
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
608 to 612nm
5% max.
Solid
42655
25g
Safety and Handling

Safety and Handling

Recommended Storage : RT