missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coomassie Brilliant Blue G-250, Fisher BioReagents

Manufacturer:  Fisher BioReagents FLBP10050

 View more versions of this product

Catalog No. BP100-50




Description

Description

Brilliant Blue G-250 is a general protein stain.
Specifications

Specifications

Brilliant Blue G-250
600g(-1) cm(-1) min.
Coomassie™ Blue G-250
RWVGQQGBQSJDQV-UHFFFAOYSA-M
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
608 to 612nm
5% max.
Solid
42655
50g
6104-58-1
C47H48N3NaO7S2
Pass Test
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
854.025
6328534
854.02
Glass Bottle
Blue

Chemical Identifiers

6104-58-1
854.025
Coomassie™ Blue G-250
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
C47H48N3NaO7S2
RWVGQQGBQSJDQV-UHFFFAOYSA-M
6328534
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents