missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brij™ L23, Alfa Aesar™

Manufacturer:  Alfa Aesar A1580930

 View more versions of this product

Catalog No. AAA1580930



Description

Description

Solubility
Soluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.
Specifications

Specifications

Brij 35
CH3(CH2)10CH2(OCH2CH2)23OH
MFCD00080891
250g
9002-92-0
CH3(CH2)10CH2(OCH2CH2)23OH
1199.57

Chemical Identifiers

9002-92-0
CH3(CH2)10CH2(OCH2CH2)23OH
MFCD00080891
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

RTECSNumber : JR5990000

TSCA : TSCA

SDS Documents