missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brij™ C20, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 344290025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brij™ C20 | |
9004-95-9 | |
Pellets | |
C56H114O21 | |
MFCD00080892 | |
10, 7449 | |
Solubility in water: soluble. Other solubilities: soluble in lower alcohols, insoluble in propylene glycol, coal-tar hydrocarbo, n, cottonseed and mineral oil | |
NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol | |
45 to 60mg KOH/g | |
1123.5 | |
Plastic bottle |
White | |
1mg KOH/g max. | |
35.4°C to 39.6°C | |
C16H33(OCH2CH2)20OH | |
2.5 kg | |
Polyoxyethylene(20) cetyl ether | |
>149°C | |
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO | |
1123.51 | |
2724259 | |
Authentic | |
1% max. |
Chemical Identifiers
9004-95-9 | |
1123.51 | |
NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
2724259 |
C56H114O21 | |
MFCD00080892 | |
Polyoxyethylene(20) cetyl ether | |
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO |
Safety and Handling
RTECSNumber : TR1581470
TSCA : TSCA
RUO – Research Use Only