missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals
$77.28 - $626.27
Chemical Identifiers
CAS | 9002-92-0 |
---|---|
Molecular Formula | C14H30O2 |
Molecular Weight (g/mol) | 230.39 |
MDL Number | MFCD00043063 |
InChI Key | SFNALCNOMXIBKG-UHFFFAOYSA-N |
Synonym | Polyoxyethylene(23)lauryl ether |
IUPAC Name | 2-(dodecyloxy)ethan-1-ol |
SMILES | CCCCCCCCCCCCOCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC228342500
|
Thermo Scientific Chemicals
228342500 |
250 g | HDPE plastic bottle |
Each for $77.28
|
|
||||
AC228345000
|
Thermo Scientific Chemicals
228345000 |
500 g | HDPE plastic bottle |
Each for $107.10
|
|
||||
AC228340010
|
Thermo Scientific Chemicals
228340010 |
1 kg | HDPE plastic bottle |
Each for $181.13
|
|
||||
AC228340051
|
Thermo Scientific Chemicals
228340051 |
5 kg | Plastic drum |
Each for $626.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-92-0 | |
230.39 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol |
C14H30O2 | |
MFCD00043063 | |
Polyoxyethylene(23)lauryl ether | |
CCCCCCCCCCCCOCCO |
Specifications
White | |
9002-92-0 | |
>100.0°C | |
C14H30O2 | |
MFCD00043063 | |
Polyoxyethylene(23)lauryl ether | |
>149°C | |
CCCCCCCCCCCCOCCO | |
40 to 60mg KOH/g | |
1199.56 | |
HDPE plastic bottle | |
Brij™ 35 |
5mg KOH/g max. | |
Waxy Pellets | |
33.0°C to 40.0°C | |
C12H25(OCH2CH2)23OH | |
250 g | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol | |
230.39 | |
Authentic | |
1% max. |
RUO – Research Use Only