Please login to your online account to display your discounted pricing

Brij 35, 30% (w/v), LabChem™

$65.66 - $65.66

Chemical Identifiers

Chemical Name or Material Brij 35
CAS 7732-18-5
CAS Max % 70
CAS 9002-92-0
CAS Max % 30
Linear Formula (C2H4O)nC12H26O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC117921

 
lab chem
LC117921
Poly Bottle 500mL Each for $65.66
Description & Specifications

Specifications

Chemical Name or Material Brij 35
Physical Form Liquid
Assay 30% (w/v)
Identification Passes Test
Linear Formula (C2H4O)nC12H26O
CAS 7732-18-5
Packaging Poly Bottle
Health Hazard 1 Danger
Solubility Soluble in water
CAS Max % 70
Health Hazard 2 GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Toxic to aquatic life.
CAS 9002-92-0
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Wear protective gloves/eye protection.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
CAS Max % 30
Recommended Storage Room Temperature
Safety and Handling

Danger

GHS P Statement
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Wear protective gloves/eye protection.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Toxic to aquatic life.

Room Temperature