missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bright Brushing Gold

$67.58 - $67.58

Chemical Identifiers

MDL Number MFCD00003436
UN Number UN1993
Chemical Name or Material Bright Brushing Gold
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1294304 SDS Alfa Aesar™
1294304
2g Each for $67.58
Specifications Description & Specifications

Chemical Identifiers

MFCD00003436
UN1993
Bright Brushing Gold

Specifications

33° (91°F)
Liquid
UN1993
Red-brown
Bright Brushing Gold
2g
MFCD00003436
Pleasant
156°C

Safety and Handling

GHS H Statement
H304-H226-H302-H315-H319-H317
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA