Please login to your online account to display your discounted pricing

Brewers yeast, ACROS Organics™


Quantity 25kg

Manufacturer: thermo fisher scientific  AC368080250

 View more versions of this product

Catalog No. AC368080250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 25kg
Color Cream to brown
Yeast and Mold 1000/g max.
Residue after Ignition 9.0% max. (on dry substance)
E. coli Absent in 1 g
Additional Information Staphylococcus aureus: absent in 1 g
MDL Number MFCD00132595
Chemical Name or Material Brewers yeast
Loss on Drying 8.0% max.
Total Nitrogen 6.5% min. (on dry substance)
Microbial Contamination 10000 /g total aerobic microbial count max.
Salmonella Absent in 25 g
CAS 68876-77-7

Specifications

Chemical Name or Material Brewers yeast
CAS 68876-77-7
MDL Number MFCD00132595