Please login to your online account to display your discounted pricing

Brewers yeast, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC368080010

 View more versions of this product

Catalog No. AC368080010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 1kg
Color Cream to brown
E. coli Absent in 1 g
Additional Information Staphylococcus aureus: absent in 1 g
Salmonella Absent in 25 g
Chemical Name or Material Brewers yeast
CAS 68876-77-7
MDL Number MFCD00132595
Loss on Drying 8.0% max.
Packaging Plastic bottle
Total Nitrogen 6.5% min. (on dry substance)
Microbial Contamination 10000 /g total aerobic microbial count max.
Yeast and Mold 1000/g max.
Residue after Ignition 9.0% max. (on dry substance)

Specifications

Chemical Name or Material Brewers yeast
CAS 68876-77-7
MDL Number MFCD00132595