Please login to your online account to display your discounted pricing

Brewers yeast, ACROS Organics™

$52.46 - $52.46

Chemical Identifiers

Chemical Name or Material Brewers yeast
CAS 68876-77-7
MDL Number MFCD00132595
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC368080010

  SDS
fisher scientific acros
AC368080010
Plastic bottle 1kg Each for $52.46

AC368080250

  SDS
thermo fisher scientific
AC368080250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC368080050

  SDS
fisher scientific acros
AC368080050
Plastic bucket 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 1kg
Color Cream to brown
E. coli Absent in 1 g
Additional Information Staphylococcus aureus: absent in 1 g
Salmonella Absent in 25 g
Chemical Name or Material Brewers yeast
CAS 68876-77-7
MDL Number MFCD00132595
Loss on Drying 8.0% max.
Packaging Plastic bottle
Total Nitrogen 6.5% min. (on dry substance)
Microbial Contamination 10000 /g total aerobic microbial count max.
Yeast and Mold 1000/g max.
Residue after Ignition 9.0% max. (on dry substance)