missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrettPhos Palladacycle 3rd Generation, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 047250MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
BrettPhos Palladacycle 3rd Generation | |
Yellow | |
250 mg | |
906.5 | |
Solid |
1470372-59-8 | |
MFCD22572666 | |
[(BrettPhos)-2-(2Â-amino-1,1Â-biphenyl)]palladium (II) methanesulfonate; BrettPhos Pd G3 | |
98% |
Chemical Identifiers
1470372-59-8 | |
MFCD22572666 | |
[(BrettPhos)-2-(2Â-amino-1,1Â-biphenyl)]palladium (II) methanesulfonate; BrettPhos Pd G3 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only