Please login to your online account to display your discounted pricing

BrdU Mouse, FITC, Clone: BU20A, eBioscience™

Mouse Monoclonal Antibody

$162.40 - $442.25

Specifications

Antigen BrdU
Clone BU20A
Host Species Murine
Gene Alias 5-bromodeoxyuridine
Species Reactivity Chimpanzee, Human, Macaque, Mouse, Non-human primate, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245239

 
life technologies
LS11507141
25 Tests Each for $162.40

50245240

 
life technologies
LS11507142
100 Tests Each for $442.25
Description & Specifications

Specifications

Antigen BrdU
Clone BU20A
Host Species Murine
Gene Alias 5-bromodeoxyuridine
Species Reactivity Chimpanzee, Human, Macaque, Mouse, Non-human primate, Rat
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

This Bu20a monoclonal antibody reacts with 5-bromodeoxyuridine (BrdU). BrdU is a derivative of uridine that can be incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Anti-BrdU can then be used to identify cells that have undergone DNA synthesis during BrdU treatment.

For staining for flow cytometric analysis, we recommend the use of the BrdU Staining Buffer Set (cat. 00-5525) and protocol.