Please login to your online account to display your discounted pricing

BrdU Mouse, Clone: BU20A, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen BrdU
Clone BU20A
Host Species Mouse
Gene Alias 5-bromodeoxyuridine
Species Reactivity Chimpanzee, Human, Macaque, Mouse, Non-human primate, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013081

 
life technologies
LS14507180
25μg Each for $85.55

5013082

 
life technologies
LS14507182
100μg Each for $158.05
Description & Specifications

Specifications

Antigen BrdU
Clone BU20A
Host Species Mouse
Gene Alias 5-bromodeoxyuridine
Species Reactivity Chimpanzee, Human, Macaque, Mouse, Non-human primate, Rat
Applications Flow Cytometry (Intracellular Staining)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This Bu20a monoclonal antibody reacts with 5-bromodeoxyuridine (BrdU). BrdU is a derivative of uridine that can be incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Anti-BrdU can then be used to identify cells that have undergone DNA synthesis during BrdU treatment.

For staining for flow cytometric analysis, we recommend the use of the BrdU Staining Buffer Set (cat. 00-5525) and protocol.