Alfa Aesar™ Brass gauze, alloy 260, 80 mesh woven from 0.14mm (0.0055 in.) dia. wire; Cu:Zn; 70:30 wt%
Manufacturer:  Alfa Aesar™ 45187GT
 View more versions of this product
Catalog No. AA45187GT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Cu:Cn 70:30 wt%
80 Mesh
Woven from 0.14mm (0.0055 in.) dia. wire
MFCD00171255
Brass gauze, Alloy 260
150 x 150mm
Odorless
Gauze
Documents