Please login to your online account to display your discounted pricing

Alfa Aesar™ Brass foil, alloy 260, 0.25mm (0.01 in.) thick, 15x45cm (5.9x18in)

Chemical Identifiers

Chemical Name or Material Brass foil, Alloy 260
Name Note 0.25mm (0.01 in.) thick, 15x45cm (5.9x18 in.)
MDL Number MFCD00171255
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13503KG

  SDS
alfa aesar™
13503KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13503KJ

  SDS
alfa aesar™
13503KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 piece
Odor Odorless
Chemical Name or Material Brass foil, Alloy 260
Name Note 0.25mm (0.01 in.) thick, 15x45cm (5.9x18 in.)
Physical Form Foil
TSCA TSCA
MDL Number MFCD00171255