missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brass Foil, 0.41mm (0.016 in.) thick, Alfa Aesar™

Form: Solid in various forms

Manufacturer:  Alfa Aesar 45160GT

 View more versions of this product

Catalog No. AA45160GT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Brass
150 x 150mm
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.546
CHEBI:30052
Yellow
Yellow foil
Odorless
MFCD00171255
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Foil, 0.41mm (0.016 in.) thick

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00171255
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
SDS Documents