Please login to your online account to display your discounted pricing

Brass Foil, 0.41mm (0.016 in.) thick, Alfa Aesar™

Form: Solid in various forms

Chemical Identifiers

Chemical Name or Material Brass
Physical Form Foil, 0.41mm (0.016 in.) thick
MDL Number MFCD00171255
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45160GT

  SDS
alfa aesar
45160GT
150 x 150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA45160HB

  SDS
alfa aesar
45160HB
300 x 300mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 150 x 150mm
Color Yellow
Chemical Name or Material Brass
Odor Odorless
Physical Form Foil, 0.41mm (0.016 in.) thick
MDL Number MFCD00171255
Appearance Yellow foil