Please login to your online account to display your discounted pricing

Brass Foil, 0.025mm (0.001 in.) thick, 15.2 x 45cm (6.0 x 18 in.), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Brass
Name Note alloy 260
Physical Form Foil, 0.025mm (0.001 in.) thick, 15.2 X 45cm (6.0 X 18 in.)
Molecular Formula Typical composition of alloy: Cu 68.5-71.5%, Pb 0.07% max , Fe 0.05% max , Zn remainder
MDL Number MFCD00171255
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13505KG

  SDS
alfa aesar
13505KG
1 Piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13505KJ

  SDS
alfa aesar
13505KJ
5 Pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Foil, 0.025mm (0.001 in.) thick, 15.2 X 45cm (6.0 X 18 in.)
Quantity 1 Piece
Color Yellow
Chemical Name or Material Brass
Odor Odorless
Name Note alloy 260
Molecular Formula Typical composition of alloy: Cu 68.5-71.5%, Pb 0.07% max , Fe 0.05% max , Zn remainder
MDL Number MFCD00171255
Appearance Yellow foil
Composition Cu 68.5 to 71.5%, Pb 0.07% max, Fe 0.05% max, Zn remainder