missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brass Foil, 0.025mm (0.001 in.) thick, 15.2 x 45cm (6.0 x 18 in.), Alfa Aesar™

Manufacturer:  Alfa Aesar 13505KJ

 View more versions of this product

Catalog No. AA13505KJ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Brass
5 pc.
Yellow foil
Cu 68.5 to 71.5%, Pb 0.07% max, Fe 0.05% max, Zn remainder
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.546
CHEBI:30052
Yellow
alloy 260
7440-50-8
Odorless
MFCD00171255
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Foil, 0.025mm (0.001 in.) thick, 15.2 X 45cm (6.0 X 18 in.)

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00171255
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Documents