Humboldt Bunsen Burner Brass Flame Spreaders
Manufacturer:  Humboldt Inc HUMH6670
 View more versions of this product
Catalog No. 03-995C
Specifications Description & Specifications
Specifications
0.68 in.
To produce a broad, flat, fan-shaped flame, in bending glass tubing and other similar applications
Brass
1.72cm
For humboldt burners equipped with flame stabilizer
Burner Wing Top