Humboldt Brass Flame Spreaders
Manufacturer:  Humboldt HUMH6650
 View more versions of this product
Catalog No. 03-995B
Description
Description
Brass Flame Spreaders. Brass wing top slips over burner tube of Bunsen burners. Makes fan-shaped flame, convenient for glass bending.
1For use with 1/2 in. (13mm) tube burners that have flame stabilizers.
Specifications
Specifications
0.5 in.
To produce a broad, flat, fan-shaped flame, in bending glass tubing and other similar applications
Brass
Burner, Wing Top
1.27 cm
For bunsen burner
Large
Documents