missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Branson Ultrasonics™ Microtips for Sonifiers SLPe, SLPt, 150E and 150T

Designed to process samples to the smallest diameter vessels.

Manufacturer:  Branson Ultrasonics™ 1091221065

 View more versions of this product

Catalog No. 15-345-122



Description

Description

 

  • Can handle volumes up to 100mL
  • Working length: 4 in.
  • Sound proof enclosure

 

Order Info

0.125 in. microtip is provided with the 150 watt units

Specifications

Specifications

0.12 in.
Ideal for processing samples to the smallest diameter vessels
10.16cm
3 to 10mL
SLP Microtip
0.32cm
4 in.
3 to 10mL
SLPe, SLPt, 150E and 150T Sonifier™
Documents