Branson Ultrasonics™ 200- and 400-Watt Sonifiers: Horns
Various horns for Branson 200- and 400-Watt Sonifiers
Manufacturer:  Branson Ultrasonics™ 611005021
 View more versions of this product
Catalog No. 15-338-57
Description
Description
For use with Branson 200- and 400-Watt Sonifiers
Tapped Horns
  • High-gain “Q” horn maintains intensity when processing with large tip diameters
  • 1/2 in. horns supplied with one replaceable tip
  • Replace only the flat tip to remedy cavitation erosion; use extender with 0.5 in. horns for environmental and other particulate samples
Cup Horns
  • Transparent polycarbonate water bath with 1/8 NPT flow ports
Specifications
Specifications
0.75 in.
200 to 500mL
Tapped Horn
1.9cm
Model 500 ultrasonic dismembrator
Documents